R&D Systems
Manufacturer Code:MAB6117SP
Catalog # MAB6117SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 549115 |
Immunogen | <EM>E. coli</EM>-derived recombinant human VAMP-7 Ala2-Met140 Accession # P51809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ54296 SYBL1 SYBL1FLJ53762 synaptobrevin-like 1 Synaptobrevin-like protein 1 tetanus neurotoxin-insensitive VAMP Tetanus-insensitive VAMP TIVAMP TI-VAMP VAMP7 VAMP-7FLJ53045 vesicle-associated membrane protein 7; synaptobrevin-like 1; Synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; Tetanus-insensitive VAMP; Ti-VAMP; VAMP-7; Vesicle-associated membrane protein 7
Gene Aliases: SYBL1; TI-VAMP; TIVAMP; VAMP-7; VAMP7
UniProt ID: (Human) P51809
Entrez Gene ID: (Human) 6845
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.