Novus Biologicals
Manufacturer Code:NBP241183
Catalog # NBP241183
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human VAMP7. The immunogen is located within amino acids 20 - 70 of VAMP7. Amino Acid Squence: AKIPSENNKLTYSHGNYL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ54296 SYBL1FLJ53762 synaptobrevin-like 1 Synaptobrevin-like protein 1 tetanus neurotoxin-insensitive VAMP Tetanus-insensitive VAMP Ti-VAMP TIVAMP VAMP-7FLJ53045 vesicle-associated membrane protein 7; synaptobrevin-like 1; Synaptobrevin-like protein 1; tetanus neurotoxin-insensitive VAMP; Tetanus-insensitive VAMP; Ti-VAMP; VAMP-7; Vesicle-associated membrane protein 7
Gene Aliases: SYBL1; TI-VAMP; TIVAMP; VAMP-7; VAMP7
UniProt ID: (Human) P51809
Entrez Gene ID: (Human) 6845
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.