Novus Biologicals
Manufacturer Code:NBP274824AF750
Catalog # NP274824A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3H5 |
Immunogen | Full length recombinant protein of human VAM1 (NP_057531) produced in E.coli. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: MAGUK p55 subfamily member 6; MAGUK protein p55T; MAGUK protein p55T membrane protein palmitoylated 6 (MAGUK p55 subfamily member 6) p55T protein associated with Lin7 2 VAM-1PALS2VAM1MAGUK p55 subfamily member 6 Veli-associated MAGUK 1; membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); protein associated with Lin7 2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: MPP6; p55T; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5
Entrez Gene ID: (Human) 51678
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.