Invitrogen
Manufacturer Code:PA587957
Catalog # PIPA587957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 220-420 of human MPP6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CH474011:EDL88200; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); P55T protein; Protein associated with Lin-7 2; protein associated with Lin7 2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: Dlgh4; MPP6; P55t; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5, (Mouse) Q9JLB0
Entrez Gene ID: (Human) 51678, (Rat) 362359, (Mouse) 56524
Molecular Function:
cell junction protein
kinase
nucleotide kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.