Invitrogen
Manufacturer Code:PA531064
Catalog # PIPA531064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 272 of Human VAM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); PALS2 VAM-1 VAM1 p55T MAGUK p55 subfamily member 6 MAGUK protein p55T VELI-associated MAGUK 1 protein associated with Lin7 2; protein associated with Lin7 2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: MPP6; p55T; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5
Entrez Gene ID: (Human) 51678
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.