Invitrogen
Manufacturer Code:PA521889
Catalog # PIPA521889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1 and 44 of VAM1 (Uniprot IDNo.Q9NZW5) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CH474011:EDL88200; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); P55T protein; PALS2 VAM-1 VAM1 p55T MAGUK p55 subfamily member 6 MAGUK protein p55T VELI-associated MAGUK 1 protein associated with Lin7 2; Protein associated with Lin-7 2; protein associated with Lin7 2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: Dlgh4; MPP6; P55t; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5, (Mouse) Q9JLB0
Entrez Gene ID: (Human) 51678, (Rat) 362359, (Mouse) 56524
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.