Novus Biologicals
Manufacturer Code:NB1001366
Catalog # NB1001366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence QQVLENLTELPSS-C corresponding to N-Terminus according to NP_057531.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MAGUK p55 subfamily member 6; MAGUK protein p55T; MAGUK protein p55T membrane protein palmitoylated 6 (MAGUK p55 subfamily member 6) p55T protein associated with Lin7 2 VAM-1PALS2VAM1MAGUK p55 subfamily member 6 Veli-associated MAGUK 1; membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); protein associated with Lin7 2; VAM-1; VELI-associated MAGUK 1
Gene Aliases: MPP6; p55T; PALS2; VAM-1; VAM1
UniProt ID: (Human) Q9NZW5
Entrez Gene ID: (Human) 51678
Molecular Function: cell junction protein kinase nucleotide kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.