Novus Biologicals
Manufacturer Code:NBP259378
Catalog # NBP259378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S6-38 |
Immunogen | Synthetic peptide amino acids 521-532 of human VAChT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC12716 solute carrier family 18 (vesicular acetylcholine) member 3 Solute carrier family 18 member 3 solute carrier family 18 member 3 VAChT VACHTvesicular acetylcholine transporter; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; Solute carrier family 18 member 3; solute carrier family 18, member 3; VAChT; Vesicular acetylcholine transporter
Gene Aliases: SLC18A3; VACHT
UniProt ID: (Human) Q16572
Entrez Gene ID: (Human) 6572
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.