Novus Biologicals
Manufacturer Code:NB100913460.1MG
Catalog # NB017900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of human mouse and rat VACHT conjugated to immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC12716 solute carrier family 18 (vesicular acetylcholine) member 3 Solute carrier family 18 member 3 solute carrier family 18 member 3 VAChT VACHTvesicular acetylcholine transporter; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; Solute carrier family 18 member 3; solute carrier family 18, member 3; VAChT; Vesicular acetylcholine transporter
Gene Aliases: SLC18A3; VACHT
UniProt ID: (Human) Q16572
Entrez Gene ID: (Human) 6572
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.