Invitrogen
Manufacturer Code:PA555685
Catalog # PIPA555685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human VAC14 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein VAC14 homolog; Protein VAC14 homolog Tax1-binding protein 2 VAC14 TAX1BP2 TRX; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; Tax1-binding protein 2; Vac14 homolog
Gene Aliases: ArPIKfyve; TAX1BP2; TRX; VAC14
UniProt ID: (Human) Q08AM6
Entrez Gene ID: (Human) 55697
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.