Novus Biologicals
Manufacturer Code:NBP305272100UL
Catalog # NB096385
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human VAC14. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ArPIKfyve FLJ10305 FLJ36622 MGC149816 protein VAC14 homolog Tax1 (human T-cell leukemia virus type I) binding protein 1 Tax1 (human T-cell leukemia virus type I) binding protein 2 Tax1-binding protein 2 TAX1BP2FLJ46582 TRXMGC149815 Vac14 homolog (S. cerevisiae); Protein VAC14 homolog; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; Tax1-binding protein 2; Vac14 homolog
Gene Aliases: ArPIKfyve; TAX1BP2; TRX; VAC14
UniProt ID: (Human) Q08AM6
Entrez Gene ID: (Human) 55697
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.