Abnova
Manufacturer Code:H00055697B01P
Catalog # 89-017-128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | VAC14 (NP_060522.3 1 a.a. ~ 782 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human VAC14 protein.
For Research Use Only
Protein Aliases: ArPIKfyve; FLJ10305; FLJ36622; FLJ46582; MGC149815; MGC149816; Protein VAC14 homolog; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; Tax1-binding protein 2; TAX1BP2; TRX; Vac14 homolog
Gene Aliases: ArPIKfyve; TAX1BP2; TRX; VAC14
UniProt ID: (Human) Q08AM6
Entrez Gene ID: (Human) 55697
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.