Proteintech
Manufacturer Code:157711AP150UL
Catalog # 50-173-0465
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VAC14 Fusion Protein Ag8415 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ArPIKfyve Protein VAC14 homolog Tax1 binding protein 2 TAX1BP2 TRX VAC14 Vac14 homolog (S. cerevisiae); Protein VAC14 homolog; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; Tax1-binding protein 2; Vac14 homolog; vacuole 14 protein; vacuole morphology 14
Gene Aliases: AA959718; ArPIKfyve; BC032215; D8Wsu151e; ingls; TAX1BP2; TRX; VAC14
UniProt ID: (Human) Q08AM6, (Rat) Q80W92, (Mouse) Q80WQ2
Entrez Gene ID: (Human) 55697, (Rat) 307842, (Mouse) 234729
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.