Novus Biologicals
Manufacturer Code:NBP2843250.1ML
Catalog # NB032253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of V1a Vasopressin R/AVPR1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1a; Antidiuretic hormone receptor 1a arginine vasopressin receptor 1A AVPR V1a AVPR1 SCCL vasopressin subtype 1a receptor V1a vasopressin receptor V1aR V1-vascular vasopressin receptor AVPR1A Vascular/hepatic-type arginine vasopressin receptor vasopressin V1a receptor; AVPR V1a; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor
Gene Aliases: AVPR V1a; AVPR1; AVPR1A; V1aR
UniProt ID: (Human) P37288
Entrez Gene ID: (Human) 552
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.