Invitrogen
Manufacturer Code:PA562681
Catalog # PIPA562681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human V-ATPase E2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6E1 ATP6EL2 ATP6V1EL2 VMA4 V-ATPase subunit E 2 V-type proton ATPase subunit E 2 vacuolar proton pump subunit E 2 vacuolar-type proton-translocating ATPase subunit E1; ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2; testis secretory sperm-binding protein Li 235P; V-ATPase subunit E 2; V-type proton ATPase subunit E 2; Vacuolar proton pump subunit E 2; vacuolar-type proton-translocating ATPase subunit E1
Gene Aliases: ATP6E1; ATP6EL2; ATP6V1E2; ATP6V1EL2; VMA4
UniProt ID: (Human) Q96A05
Entrez Gene ID: (Human) 90423
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.