Invitrogen
Manufacturer Code:PA563431
Catalog # PIPA563431
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human V-ATPase D |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6M VATD VMA8 ATPase H+ transporting lysosomal member M ATPase H+ transporting lysosomal (vacuolar proton pump) H(+)-transporting two-sector ATPase subunit M V-ATPase 28 kDa accessory protein V-ATPase D subunit V-ATPase subunit D V-type proton ATPase subunit D vac; ATPase, H+ transporting lysosomal, member M; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal 34kDa, V1 subunit D; H(+)-transporting two-sector ATPase, subunit M; V-ATPase 28 kDa accessory protein; V-ATPase D subunit; V-ATPase subunit D; V-type proton ATPase subunit D; vacuolar ATP synthase subunit D; vacuolar H-ATPase subunit D; vacuolar proton pump D subunit; vacuolar proton pump delta polypeptide; Vacuolar proton pump subunit D; vacuolar proton-ATPase subunit D
Gene Aliases: ATP6M; ATP6V1D; VATD; VMA8
UniProt ID: (Human) Q9Y5K8
Entrez Gene ID: (Human) 51382
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.