Invitrogen
Manufacturer Code:PA593258
Catalog # PIPA593258
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-280 of human ATP6V1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, V1 subunit B, isoform 1; Endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58kD subunit; H+-ATPase beta 1 subunit; lysosomal 56/58kDa; V-ATPase B1 subunit; V-ATPase subunit B 1; V-type proton ATPase subunit B, kidney isoform; vacuolar proton pump 3; Vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3
Gene Aliases: ATP6B1; ATP6V1B1; AW208839; D630003L15; D630030L16Rik; D630039P21Rik; RTA1B; VATB; VMA2; Vpp-3; VPP3
UniProt ID: (Human) P15313
Entrez Gene ID: (Human) 525, (Rat) 312488, (Mouse) 110935
Molecular Function: ATP synthase DNA binding protein anion channel cation transporter hydrolase ion channel ligand-gated ion channel nucleic acid binding receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.