Invitrogen
Manufacturer Code:PA526779
Catalog # PIPA526779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 441-468 amino acids from the central region of human ATP6V1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6A1 ATP6V1A1 HO68 VA68 VPP2 Vma1 ATPase H+ transporting lysosomal subunit A1 H(+)-transporting two-sector ATPase subunit A H+-transporting ATPase chain A vacuolar (VA68 type) V-ATPase 69 kDa subunit 1 V-ATPase A subunit 1 V-ATPase subunit A V-type proton ATPase catalytic subunit A v; ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A; ATPase, H+ transporting, lysosomal, subunit A1; H(+)-transporting two-sector ATPase, subunit A; H+-transporting ATPase chain A, vacuolar (VA68 type); V-ATPase 69 kDa subunit; V-ATPase 69 kDa subunit 1; V-ATPase A subunit 1; V-ATPase subunit A; V-type proton ATPase catalytic subunit A; vacuolar ATP synthase catalytic subunit A, ubiquitous isoform; Vacuolar ATPase isoform VA68; vacuolar proton pump alpha subunit 1; Vacuolar proton pump subunit alpha
Gene Aliases: ATP6A1; ATP6V1A; ATP6V1A1; HO68; VA68; Vma1; VPP2
UniProt ID: (Human) P38606
Entrez Gene ID: (Human) 523
Molecular Function:
ATP synthase
DNA binding protein
anion channel
cation transporter
hydrolase
ion channel
ligand-gated ion channel
nucleic acid binding
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.