Novus Biologicals
Manufacturer Code:NBP289650G
Catalog # NB067564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 135 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Uteroglobin/SCGB1A1 (P11684 Met1-Asn91). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: blastokinin; blastokinin CC10Urine protein 1 CC16 CCSPCCPBP Clara cell phospholipid-binding protein Clara cells 10 kDa secretory protein Secretoglobin family 1A member 1 secretoglobin family 1A member 1 (uteroglobin) UGBUP-1 UP1 Urinary protein 1 uteroglobin; CC10; CCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; Urinary protein 1; urine protein 1; Uteroglobin
Gene Aliases: CC10; CC16; CCPBP; CCSP; SCGB1A1; UGB; UP-1; UP1
UniProt ID: (Human) P11684
Entrez Gene ID: (Human) 7356
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.