R&D Systems
Manufacturer Code:AF4218SP
Catalog # AF4218SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>E. coli</em>-derived recombinant human Uteroglobin/SCGB1A1 Glu22-Asn91 Accession # NP_003348 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: blastokinin; blastokinin CC10 CC10Urine protein 1 CC16 CCPBP CCSP CCSPCCPBP Clara Clara cell phospholipid-binding protein Clara cells 10 kDa secretory protein PCB-BP SCGB1A1 Secretoglobin family 1A member 1 secretoglobin family 1A member 1 (uteroglobin) UGB UGBUP-1 UP1 Urinary protein 1 uteroglobin; CC10; CCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; Urinary protein 1; urine protein 1; Uteroglobin
Gene Aliases: CC10; CC16; CCPBP; CCSP; SCGB1A1; UGB; UP-1; UP1
UniProt ID: (Human) P11684
Entrez Gene ID: (Human) 7356
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.