Invitrogen
Manufacturer Code:MA529786
Catalog # PIMA529786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 30 |
Immunogen | Recombit Human Uteroglobin/SCGB1A1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: blastokinin; CC10; CCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; SCGB1A1; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; Urinary protein 1; urine protein 1; Uteroglobin
Gene Aliases: CC10; CC16; CCPBP; CCSP; SCGB1A1; UGB; UP-1; UP1
UniProt ID: (Human) P11684
Entrez Gene ID: (Human) 7356
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.