Invitrogen
Manufacturer Code:MA529788
Catalog # PIMA529788
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 27 |
Immunogen | Recombit Mouse Uteroglobin/SCGB1A1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Blastokinin; CC10; CCPBP; Clara cell 17 kDa protein; Clara cell phospholipid-binding protein; clara cell secretory protein; Clara cells 10 kDa secretory protein; PCB-binding protein; SCGB1A1; Secretoglobin family 1A member 1; Uteroglobin
Gene Aliases: CC10; CC16; CCSP; PCB-BP; Scgb1a1; UG; UGB; Utg
UniProt ID: (Mouse) Q06318
Entrez Gene ID: (Mouse) 22287
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.