Invitrogen
Manufacturer Code:MA535996
Catalog # PIMA535996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 135 |
Immunogen | Recombinant Human Uteroglobin/SCGB1A1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: blastokinin; Blastokinin CC10 CC16 CCPBP CCSP Clara cell 17 kDa protein Clara cell phospholipid-binding protein clara cell secretory protein Clara cells 10 kDa secretory protein PCBB PCB-binding protein PCB-BP Scgb1a1 Secretoglobin family 1A member 1 secretoglobin family 1A member 1 (uteroglobin) UG Ugb UP1 UP-1 Urinary protein 1 urine protein 1 Uteroglobin Uteroglobin (Clara cell secretory protein) Utg; CC10; CCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; Urinary protein 1; urine protein 1; Uteroglobin
Gene Aliases: CC10; CC16; CCPBP; CCSP; SCGB1A1; UGB; UP-1; UP1
UniProt ID: (Human) P11684
Entrez Gene ID: (Human) 7356
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.