Invitrogen
Manufacturer Code:PA579957
Catalog # PIPA579957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived rat Uteroglobin recombit protein (Position: S20-V96). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Blastokinin; CC10; CCPBP; Clara cell 17 kDa protein; Clara cell phospholipid-binding protein; clara cell secretory protein; Clara cells 10 kDa secretory protein; PCB-binding protein; Secretoglobin family 1A member 1; Uteroglobin; Uteroglobin (Clara cell secretory protein); Uteroglobin Clara cell phospholipid-binding protein CCPBP Clara cells 10 kDa secretory protein CC10 PCB-binding protein Secretoglobin family 1A member 1 Scgb1a1 Cc10 Ugb Utg
Gene Aliases: CC10; CC16; CCSP; PCB-BP; PCBB; Scgb1a1; UG; UGB; Utg
UniProt ID: (Mouse) Q06318, (Rat) P17559
Entrez Gene ID: (Mouse) 22287, (Rat) 25575
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.