Invitrogen
Manufacturer Code:PA578215
Catalog # PIPA578215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center of human Uteroglobin. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: blastokinin; CC10; CCPBP; Clara cell phospholipid-binding protein; Clara cells 10 kDa secretory protein; PCB-binding protein; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; UP1 CCSP secretoglobin family 1A member 1 (uteroglobin) UP-1 CCPBP SCGB1A1 CC10 CC16 UGB; Urinary protein 1; urine protein 1; Uteroglobin; Uteroglobin (Clara cell secretory protein)
Gene Aliases: CC10; CC16; CCPBP; CCSP; PCBB; SCGB1A1; UGB; UP-1; UP1; Utg
UniProt ID: (Human) P11684, (Rat) P17559
Entrez Gene ID: (Human) 7356, (Rat) 25575
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.