Proteintech
Manufacturer Code:104901AP150UL
Catalog # 50-172-6347
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Uteroglobin Fusion Protein Ag0758 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Blastokinin; CC10; CC10 CC16 CCPBP CCSP SCGB1A1 UGB UP 1 UP1 Urinary protein 1 Urine protein 1 Uteroglobin; CCPBP; Clara cell 17 kDa protein; Clara cell phospholipid-binding protein; clara cell secretory protein; Clara cells 10 kDa secretory protein; PCB-binding protein; Secretoglobin family 1A member 1; secretoglobin, family 1A, member 1 (uteroglobin); UP-1; Urinary protein 1; urine protein 1; Uteroglobin
Gene Aliases: CC10; CC16; CCPBP; CCSP; PCB-BP; SCGB1A1; UG; UGB; UP-1; UP1; Utg
UniProt ID: (Human) P11684, (Mouse) Q06318
Entrez Gene ID: (Human) 7356, (Mouse) 22287
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.