Abnova
Manufacturer Code:PAB11846
Catalog # 89-115-421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 327-346 of mouse Usf2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of Usf2.
For Research Use Only
Protein Aliases: Major late transcription factor 2; MGC91056; Upstream stimulatory factor 2; Upstream transcription factor 2; Usf-2
Gene Aliases: bHLHb12; Usf-2; Usf2
UniProt ID: (Mouse) Q64705
Entrez Gene ID: (Mouse) 22282
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.