Invitrogen
Manufacturer Code:PA534300
Catalog # PIPA534300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 1st cytoplasmic domain of human UTS2R / GPR14. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 14; G-protein coupled receptor 14; UR-2-R; UR-II-R; Urotensin II receptor; Urotensin-2 receptor; UTS2R G protein-coupled receptor 14 GPR14 G-protein coupled receptor 14 UII-receptor Urotensin 2 receptor UT-II UTR UTR2 Uii-r UR-2-R Senr Uii-r1a Uiir UR-II-R Urotensin II receptor Urotensin-2 receptor
Gene Aliases: GPR14; UR-2-R; UTR; UTR2; UTS2R
UniProt ID: (Human) Q9UKP6
Entrez Gene ID: (Human) 2837
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.