R&D Systems
Manufacturer Code:FAB9245G100UG
Catalog # FAB9245G100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 443707 |
Immunogen | Mouse Urotensin-II R synthetic peptide |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Rat MonoclonalAntibody
For Research Use Only
Protein Aliases: G protein-coupled receptor 14; G protein-coupled receptor 14 GPR14 G-protein coupled receptor 14 UR-2-R UR-II-R urotensin 2 receptor urotensin II receptor urotensin-2 receptor UrotensinIIR Urotensin-IIR UTR UTR2 UTS2R; G-protein coupled receptor 14; UR-2-R; UR-II-R; Urotensin II receptor; Urotensin-2 receptor
Gene Aliases: GPR14; UR-2-R; UTR; UTR2; UTS2R
UniProt ID: (Human) Q9UKP6
Entrez Gene ID: (Human) 2837
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.