Novus Biologicals
Manufacturer Code:NBP29414020UL
Catalog # NB022838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 35-124 of human UTS2 (NP_006777.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: prepro U-II; U-II; UCN2 UIIPRO1068 U-IIUrotensin II urotensin 2 urotensin-2; Urotensin II; Urotensin-2
Gene Aliases: PRO1068; U-II; UCN2; UII; UNQ525/PRO1068; UTS2
UniProt ID: (Human) Q6UXF6
Entrez Gene ID: (Human) 10911
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.