Novus Biologicals
Manufacturer Code:NBP294533100UL
Catalog # NB023590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 19-207 of human UPK3A (NP_008884.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC119178 UP3a UPIII UPIIIA UPK3 UPK3A uroplakin 3 uroplakin 3A Uroplakin III uroplakin3a uroplakin-3a; UP3a; UPIII; uroplakin 3; Uroplakin III; Uroplakin-3a
Gene Aliases: UP3A; UPIII; UPIIIA; UPK3; UPK3A
UniProt ID: (Human) O75631
Entrez Gene ID: (Human) 7380
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.