Invitrogen
Manufacturer Code:PA546959
Catalog # PIPA546959
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Uromodulin isoform 1 Asp25-Ser614 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Tamm-Horsfall urinary glycoprotein; THP; UMOD ADMCKD2 FJHN HNFJ HNFJ1 MCKD2 THGP THP Tamm-Horsfall glycoprotein tamm-Horsfall urinary glycoprotein uromodulin uromucoid Tamm-Horsfall glycoprotein uromucoid; Uromodulin; Uromodulin, secreted form; uromucoid
Gene Aliases: ADMCKD2; FJHN; HNFJ; HNFJ1; MCKD2; THGP; THP; UMOD
UniProt ID: (Human) P07911
Entrez Gene ID: (Human) 7369
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.