Proteintech
Manufacturer Code:119111AP150UL
Catalog # 50-553-754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Uromodulin Fusion Protein Ag2494 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADMCKD2 FJHN HNFJ MCKD2 THGP THP UMOD uromodulin; Tamm-Horsfall urinary glycoprotein; THP; Urmodulin (Tamm-Horsfall protein); Uromodulin; Uromodulin, secreted form; uromucoid
Gene Aliases: ADMCKD2; FJHN; HNFJ; HNFJ1; MCKD2; THGP; THP; UMOD; Urehd1; urehr4
UniProt ID: (Human) P07911, (Rat) P27590, (Mouse) Q91X17
Entrez Gene ID: (Human) 7369, (Rat) 25128, (Mouse) 22242
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.