Invitrogen
Manufacturer Code:MA531101
Catalog # PIMA531101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D3F2B11 |
Immunogen | Recombit Human uPA protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATF UPA URK u-PA; plasminogen activator, urinary; U-plasminogen activator; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A
Gene Aliases: ATF; BDPLT5; PLAU; QPD; u-PA; UPA; URK
UniProt ID: (Human) P00749
Entrez Gene ID: (Human) 5328
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.