Invitrogen
Manufacturer Code:PA595504
Catalog # PIPA595504
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human PLAU recombit protein (Position: I179-L431). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: plasminogen activator, urinary; U-plasminogen activator; uPA; Urinary plasminogen activator, urokinase; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A; Urokinase-type plasminogen activator U-plasminogen activator uPA Urokinase-type plasminogen activator long chain A Urokinase-type plasminogen activator short chain A Urokinase-type plasminogen activator chain B PLAU
Gene Aliases: ATF; BDPLT5; PLAU; QPD; u-PA; UPA; UPAM; URK
UniProt ID: (Human) P00749, (Mouse) P06869
Entrez Gene ID: (Human) 5328, (Rat) 25619, (Mouse) 18792
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.