Novus Biologicals
Manufacturer Code:NBP262804
Catalog # NBP262804
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
In Situ Hybridization (ISH) (ISH) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2D7 |
Immunogen | IMAC-purified SUMO-eXact-preproUCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4degreesC. |
Mouse Monoclonal Antibody
Protein Aliases: MGC129974 MGC129975 urocortin urocortin preproprotein UROCUI; prepro-urocortin; Urocortin; urocortin, preproprotein
Gene Aliases: UCN; UI; UROC
UniProt ID: (Human) P55089
Entrez Gene ID: (Human) 7349
Molecular Function:
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.