Invitrogen
Manufacturer Code:PA538248
Catalog # PIPA538248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human urocortin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Corticotensin; prepro-urocortin; UI UROC urocortin preproprotein; Urocortin; urocortin, preproprotein
Gene Aliases: Mpv17; UCN; Ucn1; UI; UROC
UniProt ID: (Human) P55089, (Rat) P55090, (Mouse) P81615
Entrez Gene ID: (Human) 7349, (Rat) 29151, (Mouse) 22226
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.