Proteintech
Manufacturer Code:262011AP150UL
Catalog # 50-173-5253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Unc119b Fusion Protein Ag24599 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA407659 AI414892 unc 119 homolog B (C. elegans) Unc119b; POC7 centriolar protein homolog B; Protein unc-119 homolog B; unc-119 homolog B
Gene Aliases: AA407659; AI414892; POC7B; UNC119B
UniProt ID: (Human) A6NIH7, (Mouse) Q8C4B4
Entrez Gene ID: (Human) 84747, (Dog) 611530, (Mouse) 106840
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.