Abnova
Manufacturer Code:MAB8723
Catalog # 89-113-542
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to N-terminus residues of zebrafish uhrf1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant uhrf1.
For Research Use Only
Protein Aliases: E3 ubiquitin-protein ligase UHRF1; fb97f09; NP95; nuclear protein; RING-type E3 ubiquitin transferase UHRF1; Ubiquitin-like PHD and RING finger domain-containing protein 1; ubiquitin-like, containing PHD and RING finger domains, 1; Ubiquitin-like-containing PHD and RING finger domains protein 1; unm b1115; wu:fb97f09; zgc:63539
Gene Aliases: fb97f09; NP95; uhrf1; unm_b1115; wu:fb97f09; zgc:63539
UniProt ID: (Zebrafish) E7EZF3
Entrez Gene ID: (Zebrafish) 406350
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.