Invitrogen
Manufacturer Code:PA576144
Catalog # PIPA576144
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide of human Poly-Ubiquitin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: polyubiquitin C; Polyubiquitin-C; UBC HMG20 Ubiquitin; Ubiquitin; Ubiquitin-related; Ubiquitin-related 1; Ubiquitin-related 2
Gene Aliases: 2700054O04Rik; AI194771; HMG20; Rps27a; TI-225; Uba52; Ubb; UBC
UniProt ID: (Human) Q9BX98, (Mouse) P0CG50, (Rat) Q63429
Entrez Gene ID: (Human) 7316, (Mouse) 22190, (Rat) 50522
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.