Invitrogen
Manufacturer Code:MA535382
Catalog # PIMA535382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0811 |
Immunogen | A synthesized peptide derived from human K48-linkage Specific Ubiquitin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AL033289 HEL-S-50 HUBCEP52 im:6892314 MGC127041 Polyubiquitin-B RPL40 Rps27a si:ch211-202a12.3 Uba52 Ubb Ubb2 Ubc Ubi-p63E Ubiquitin Ubiquitin B Ubiquitin-related zgc:172187 zUBC; epididymis secretory protein Li 50; polyubiquitin B; Polyubiquitin-B; Ubiquitin
Gene Aliases: HEL-S-50; UBB
UniProt ID: (Human) P0CG47
Entrez Gene ID: (Human) 7314
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.