R&D Systems
Manufacturer Code:MAB7031SP
Catalog # MAB7031SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 83456 |
Immunogen | <em>E. coli</em>-derived recombinant human Ubiquitin+1 Met1-Gly75 YADLREDPDRQDHHPGSGAQ (C-terminal sequence generated by a frameshift) Accession # CAA44911 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 50; FLJ25987 MGC8385 polyubiquitin B polyubiquitin-B RPS27A UBA52 UBC ubiquitin B; polyubiquitin B; Polyubiquitin-B; Ubiquitin
Gene Aliases: HEL-S-50; UBB
UniProt ID: (Human) P0CG47
Entrez Gene ID: (Human) 7314
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.