Invitrogen
Manufacturer Code:MA517196
Catalog # PIMA517196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3C12 |
Immunogen | Purified recombit fragment of human UBB expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 50; polyubiquitin B; Polyubiquitin-B; Ubiquitin
Gene Aliases: AL033289; HEL-S-50; Rps27a; Uba52; UBB; Ubb2; Ubc
UniProt ID: (Human) P0CG47, (Mouse) Q8VCH1
Entrez Gene ID: (Human) 7314, (Mouse) 22187
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.