Invitrogen
Manufacturer Code:PA570602
Catalog # PIPA570602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Ubiquitin B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L40 HUBCEP52 RPL40 Ubiquitin 52 amino acid fusion protein Ubiquitin A52 residue ribosomal protein fusion product 1 UBB UBC; epididymis secretory protein Li 50; polyubiquitin; polyubiquitin B; Polyubiquitin-B; Ubiquitin; ubiquitin C; Ubiquitin-related; zUBC
Gene Aliases: AL033289; HEL-S-50; im:6892314; Rps27a; si:ch211-202a12.3; Uba52; UBB; Ubb2; UBC; Ubi-p63E; zgc:172187; zUBC
UniProt ID: (Human) P0CG47, (Sheep) P0CG55, (Mouse) Q8VCH1
Entrez Gene ID: (Rabbit) 100351347, (Human) 7314, (Sheep) 443016, (Guinea pig) 100722422, (Mouse) 22187, (Rat) 192255, (Zebrafish) 550134
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.