Novus Biologicals
Manufacturer Code:NBP198436
Catalog # NBP198436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is ubiquitin B - N-terminal region. Peptide sequence KEGIPPDQQRLIFAGKQLEDGRTLSDYNIQKESTLHLVLRLRGGMQIFVK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis secretory protein Li 50; FLJ25987 MGC8385 polyubiquitin B polyubiquitin-B RPS27A UBA52 UBC ubiquitin B; polyubiquitin B; Polyubiquitin-B; Ubiquitin
Gene Aliases: HEL-S-50; UBB
UniProt ID: (Human) P0CG47
Entrez Gene ID: (Human) 7314
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.