Invitrogen
Manufacturer Code:LFMA0118
Catalog # AML0118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A1 |
Immunogen | Purified recombit human ubiquitin GST-fusion protein expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L40 HUBCEP52 RPL40 Ubiquitin 52 amino acid fusion protein Ubiquitin A52 residue ribosomal protein fusion product 1 Ubiquiti; epididymis secretory protein Li 50; polyubiquitin B; Polyubiquitin-B; Ubiquitin
Gene Aliases: HEL-S-50; UBB
UniProt ID: (Human) P0CG47
Entrez Gene ID: (Human) 7314
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.