R&D Systems
Manufacturer Code:MAB8595SP
Catalog # MAB8595SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1002A |
Immunogen | HEK293 human embryonic kidney cell line transfected with human Ubiquitin Accession # P0CG47 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S27a; epididymis luminal protein 112; RPS27A UBA52 UBB UBB ubiquitin B UBC; Small ribosomal subunit protein eS31; Ubiquitin; ubiquitin and ribosomal protein S27a; ubiquitin C; Ubiquitin carboxyl extension protein 80; Ubiquitin-40S ribosomal protein S27a; ubiquitin-CEP80
Gene Aliases: CEP80; HEL112; RPS27A; S27A; UBA80; UBC; UBCEP1; UBCEP80
UniProt ID: (Human) P62979
Entrez Gene ID: (Human) 6233
Molecular Function:
RNA binding protein
nucleic acid binding
ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.