R&D Systems
Manufacturer Code:A101
Catalog # A101RD
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1001C |
Immunogen | K48-linkage from human ubiquitin Accession No. P0CG47 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 12 months from date of receipt -20°C as supplied. 3 months -20°C under sterile conditions after opening. |
Rabbit Monoclonal Antibody
Protein Aliases: 40S ribosomal protein S27a; epididymis luminal protein 112; RPS27A UBA52 UBB UBB ubiquitin B UBC; Small ribosomal subunit protein eS31; Ubiquitin; ubiquitin and ribosomal protein S27a; ubiquitin C; Ubiquitin carboxyl extension protein 80; Ubiquitin-40S ribosomal protein S27a; ubiquitin-CEP80
Gene Aliases: CEP80; HEL112; RPS27A; S27A; UBA80; UBC; UBCEP1; UBCEP80
UniProt ID: (Human) P62979
Entrez Gene ID: (Human) 6233
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.