Invitrogen
Manufacturer Code:PA516829
Catalog # PIPA516829
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full-length human ubiquitin protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 40S ribosomal protein S27a; 40S ribosomal protein S27aubiquitin and ribosomal protein S27a ubiquitin carboxyl extension protein 80 ubiquitin-CEP80; epididymis luminal protein 112; Small ribosomal subunit protein eS31; Ubiquitin; ubiquitin and ribosomal protein S27a; ubiquitin C; Ubiquitin carboxyl extension protein 80; Ubiquitin-40S ribosomal protein S27a; ubiquitin-40S ribosomal protein S27a-like; ubiquitin-CEP80
Gene Aliases: 0610006J14Rik; CEP80; HEL112; RPS27A; S27A; Uba52; UBA80; Ubb; UBC; UBCEP1; UBCEP80
UniProt ID: (Rat) P62982, (Human) P62979, (Mouse) P49664
Entrez Gene ID: (Rat) 100912032, (Human) 6233, (Mouse) 78294
Molecular Function: RNA binding protein nucleic acid binding ribosomal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.