Novus Biologicals
Manufacturer Code:NBP293303100UL
Catalog # NB028491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human UBQLN2 (NP_038472.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chap1; Chap1 CHAP1/DSK2 DSK2 DSK2 homolog FLJ10167 FLJ56541 hPLIC-2 N4BP4LIC-2 Nedd4 binding protein 4 PLIC-2 PLIC2Dsk2 Protein linking IAP with cytoskeleton 2 RIHFB2157 ubiquilin 2 ubiquilin-2 Ubiquitin-like product Chap1/Dsk2; DSK2 homolog; Nedd4 binding protein 4; PLIC-2; Protein linking IAP with cytoskeleton 2; Ubiquilin-2; Ubiquitin-like product Chap1/Dsk2
Gene Aliases: ALS15; CHAP1; DSK2; HRIHFB2157; N4BP4; PLIC2; UBQLN2
UniProt ID: (Human) Q9UHD9
Entrez Gene ID: (Human) 29978
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.